missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-N,N-dicyclohexylbenzamide, 97%

$211.27 - $675.45

Chemical Identifiers

CAS 349089-31-2
Molecular Formula C19H26ClNO
Molecular Weight (g/mol) 319.873
MDL Number MFCD00540918
InChI Key BMQOLGWVHWPNTP-UHFFFAOYSA-N
PubChem CID 2189789
IUPAC Name 4-chloro-N,N-dicyclohexylbenzamide
SMILES C1CCC(CC1)N(C2CCCCC2)C(=O)C3=CC=C(C=C3)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59886MD View Documents Alfa Aesar™
H59886MD
250mg Each for $211.27
 
AAH5988603 View Documents Alfa Aesar™
H5988603
1g Each for $675.45
 
Specifications

Chemical Identifiers

349089-31-2
319.873
BMQOLGWVHWPNTP-UHFFFAOYSA-N
4-chloro-N,N-dicyclohexylbenzamide
C19H26ClNO
MFCD00540918
2189789
C1CCC(CC1)N(C2CCCCC2)C(=O)C3=CC=C(C=C3)Cl

Specifications

250mg
C19H26ClNO
BMQOLGWVHWPNTP-UHFFFAOYSA-N
4-chloro-N,N-dicyclohexylbenzamide
2189789
97%
349089-31-2
MFCD00540918
C1CCC(CC1)N(C2CCCCC2)C(=O)C3=CC=C(C=C3)Cl
319.873
319.87
4-Chloro-N,N-dicyclohexylbenzamide