missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-N-ethylbenzamide, 97%

Manufacturer:  Alfa Aesar™ H5907203

 View more versions of this product

Catalog No. AAH5907203



Description

Description

Applications
It is used as pharmaceutical intermediate.

Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications

Specifications

4-Chloro-N-ethylbenzamide
26930-17-6
MFCD00871156
TXRYLYLDCHDRSJ-UHFFFAOYSA-N
4-chloro-N-ethylbenzamide
101292
Solid
1g
C9H10ClNO
benzamide, 4-chloro-n-ethyl, 4-chloro-n-ethyl-benzamide, n-ethyl-4-chlorobenzamide, 4-chlorobenzamide, n-ethyl, benzamide, p-chloro-n-ethyl, 4-chloro-n-ethylbenzenecarboxamide, 4-chlorophenyl-n-ethylcarboxamide
CCNC(=O)C1=CC=C(C=C1)Cl
183.635
183.64
97%

Chemical Identifiers

26930-17-6
183.635
TXRYLYLDCHDRSJ-UHFFFAOYSA-N
101292
CCNC(=O)C1=CC=C(C=C1)Cl
C9H10ClNO
MFCD00871156
benzamide, 4-chloro-n-ethyl, 4-chloro-n-ethyl-benzamide, n-ethyl-4-chlorobenzamide, 4-chlorobenzamide, n-ethyl, benzamide, p-chloro-n-ethyl, 4-chloro-n-ethylbenzenecarboxamide, 4-chlorophenyl-n-ethylcarboxamide
4-chloro-N-ethylbenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents