missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-N-cyclohexylbenzamide, 97%

$226.85 - $725.91

Chemical Identifiers

CAS 57707-20-7
Molecular Formula C13H16ClNO
Molecular Weight (g/mol) 237.727
MDL Number MFCD00029412
InChI Key KSXFAESIEUQWLY-UHFFFAOYSA-N
Synonym 4-chloro-n~1~-cyclohexylbenzamide
PubChem CID 225461
IUPAC Name 4-chloro-N-cyclohexylbenzamide
SMILES C1CCC(CC1)NC(=O)C2=CC=C(C=C2)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55146MD View Documents Alfa Aesar™
H55146MD
250mg Each for $226.85
 
AAH5514603 View Documents Alfa Aesar™
H5514603
1g Each for $725.91
 
Specifications

Chemical Identifiers

57707-20-7
237.727
KSXFAESIEUQWLY-UHFFFAOYSA-N
225461
C1CCC(CC1)NC(=O)C2=CC=C(C=C2)Cl
C13H16ClNO
MFCD00029412
4-chloro-n~1~-cyclohexylbenzamide
4-chloro-N-cyclohexylbenzamide

Specifications

250mg
C13H16ClNO
4-chloro-n~1~-cyclohexylbenzamide
C1CCC(CC1)NC(=O)C2=CC=C(C=C2)Cl
237.727
237.73
4-Chloro-N-cyclohexylbenzamide
57707-20-7
MFCD00029412
KSXFAESIEUQWLY-UHFFFAOYSA-N
4-chloro-N-cyclohexylbenzamide
225461
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures