missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-N-cyclohexylbenzamide, 97%

Manufacturer:  Alfa Aesar™ H55146MD

 View more versions of this product

Catalog No. AAH55146MD



Specifications

Specifications

4-Chloro-N-cyclohexylbenzamide
57707-20-7
MFCD00029412
KSXFAESIEUQWLY-UHFFFAOYSA-N
4-chloro-N-cyclohexylbenzamide
225461
97%
250mg
C13H16ClNO
4-chloro-n~1~-cyclohexylbenzamide
C1CCC(CC1)NC(=O)C2=CC=C(C=C2)Cl
237.727
237.73

Chemical Identifiers

57707-20-7
237.727
KSXFAESIEUQWLY-UHFFFAOYSA-N
225461
C1CCC(CC1)NC(=O)C2=CC=C(C=C2)Cl
C13H16ClNO
MFCD00029412
4-chloro-n~1~-cyclohexylbenzamide
4-chloro-N-cyclohexylbenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents