missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-N-cyclohexyl-3-nitrobenzamide, 97%

Chemical Identifiers

CAS 321531-75-3
Molecular Formula C13H15ClN2O3
Molecular Weight (g/mol) 282.724
MDL Number MFCD01182372
InChI Key NXILMCJKLLFJES-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrophenyl-n-cyclohexylcarboxamide
PubChem CID 711288
IUPAC Name 4-chloro-N-cyclohexyl-3-nitrobenzamide
SMILES C1CCC(CC1)NC(=O)C2=CC(=C(C=C2)Cl)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59836MD View Documents Alfa Aesar™
H59836MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5983603 View Documents Alfa Aesar™
H5983603
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

321531-75-3
282.724
NXILMCJKLLFJES-UHFFFAOYSA-N
711288
C1CCC(CC1)NC(=O)C2=CC(=C(C=C2)Cl)[N+](=O)[O-]
C13H15ClN2O3
MFCD01182372
4-chloro-3-nitrophenyl-n-cyclohexylcarboxamide
4-chloro-N-cyclohexyl-3-nitrobenzamide

Specifications

250mg
C13H15ClN2O3
4-chloro-3-nitrophenyl-n-cyclohexylcarboxamide
C1CCC(CC1)NC(=O)C2=CC(=C(C=C2)Cl)[N+](=O)[O-]
282.724
282.72
4-Chloro-N-cyclohexyl-3-nitrobenzamide
321531-75-3
MFCD01182372
NXILMCJKLLFJES-UHFFFAOYSA-N
4-chloro-N-cyclohexyl-3-nitrobenzamide
711288
97%