missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-N-(3-methylphenyl)butyramide, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H58249MC

 View more versions of this product

Catalog No. AAH58249MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Chloro-N-(3-methylphenyl)butyramide
C11H14ClNO
100mg
GBYXVHYDSWAERN-UHFFFAOYSA-N
4-chloro-N-(3-methylphenyl)butanamide
1809759
97%
73863-44-2
MFCD00086969
4-chloro-n-3-methylphenyl butanamide, butanamide, 4-chloro-n-3-methylphenyl, 4-chloro-n-m-tolyl butanamide, n-4-chlorobutyryl-m-toluidine, 4-chloro-meta-butyrotoluidide, 4-chloro-n-3-methylphenyl butyramide
CC1=CC=CC(NC(=O)CCCCl)=C1
211.69
211.69

Chemical Identifiers

73863-44-2
211.69
GBYXVHYDSWAERN-UHFFFAOYSA-N
1809759
CC1=CC=CC(NC(=O)CCCCl)=C1
C11H14ClNO
MFCD00086969
4-chloro-n-3-methylphenyl butanamide, butanamide, 4-chloro-n-3-methylphenyl, 4-chloro-n-m-tolyl butanamide, n-4-chlorobutyryl-m-toluidine, 4-chloro-meta-butyrotoluidide, 4-chloro-n-3-methylphenyl butyramide
4-chloro-N-(3-methylphenyl)butanamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions