missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-N-(2-fluorobenzyl)aniline, 97%

Manufacturer:  Alfa Aesar™ H5627503

 View more versions of this product

Catalog No. AAH5627503



Specifications

Specifications

4-Chloro-N-(2-fluorobenzyl)aniline
723753-84-2
MFCD05740323
QCLJXZCWBRKJHD-UHFFFAOYSA-N
4-chloro-N-[(2-fluorophenyl)methyl]aniline
834588
97%
1g
C13H11ClFN
4-chloro-n-2-fluorophenyl methyl aniline, n-4-chlorophenyl-n-2-fluorobenzyl amine, 4-chloro-n-2-fluorobenzyl aniline
C1=CC=C(C(=C1)CNC2=CC=C(C=C2)Cl)F
235.686
235.69

Chemical Identifiers

723753-84-2
235.686
QCLJXZCWBRKJHD-UHFFFAOYSA-N
834588
C1=CC=C(C(=C1)CNC2=CC=C(C=C2)Cl)F
C13H11ClFN
MFCD05740323
4-chloro-n-2-fluorophenyl methyl aniline, n-4-chlorophenyl-n-2-fluorobenzyl amine, 4-chloro-n-2-fluorobenzyl aniline
4-chloro-N-[(2-fluorophenyl)methyl]aniline
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents