missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide, 97%, Alfa Aesar™

$56.58 - $668.59

Chemical Identifiers

CAS 41859-57-8
Molecular Formula C15H14ClNO2
Molecular Weight (g/mol) 275.732
MDL Number MFCD00218513
InChI Key ZTLWJYCDAXUIBK-UHFFFAOYSA-N
Synonym 4-chloro-n-2-4-hydroxyphenyl ethyl benzamide
PubChem CID 198234
IUPAC Name 4-chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide
SMILES C1=CC(=CC=C1CCNC(=O)C2=CC=C(C=C2)Cl)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54344MD View Documents Thermo Fisher Scientific
H54344MD
250mg Each for $56.58
 
AAH5434403 View Documents Thermo Fisher Scientific
H5434403
1g Each for $168.76
 
AAH5434406 View Documents Thermo Fisher Scientific
H5434406
5g Each for $668.59
 
Specifications

Chemical Identifiers

41859-57-8
275.732
ZTLWJYCDAXUIBK-UHFFFAOYSA-N
198234
C1=CC(=CC=C1CCNC(=O)C2=CC=C(C=C2)Cl)O
C15H14ClNO2
MFCD00218513
4-chloro-n-2-4-hydroxyphenyl ethyl benzamide
4-chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide

Specifications

250mg
C15H14ClNO2
4-chloro-n-2-4-hydroxyphenyl ethyl benzamide
C1=CC(=CC=C1CCNC(=O)C2=CC=C(C=C2)Cl)O
275.732
275.73
4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide
41859-57-8
MFCD00218513
ZTLWJYCDAXUIBK-UHFFFAOYSA-N
4-chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide
198234
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 255-566-3

RTECSNumber : CV2450481

TSCA : No

Recommended Storage : Ambient temperatures