missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5434406

 View more versions of this product

Catalog No. AAH5434406



Description

Description

Specifications

Specifications

4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide
41859-57-8
MFCD00218513
ZTLWJYCDAXUIBK-UHFFFAOYSA-N
4-chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide
198234
97%
5g
C15H14ClNO2
4-chloro-n-2-4-hydroxyphenyl ethyl benzamide, 4-chloro-n-4-hydroxyphenethyl benzamide, benzamide, 4-chloro-n-2-4-hydroxyphenyl ethyl, benzamide, p-chloro-n-2-p-hydroxyphenyl ethyl, bez-da, n-4-chlorobenzoyl tyramine, n-4-chlorobenzoyl-tyramine, p-chloro-n-2-p-hydroxyphenyl ethyl benzamide
C1=CC(=CC=C1CCNC(=O)C2=CC=C(C=C2)Cl)O
275.732
275.73

Chemical Identifiers

41859-57-8
275.732
ZTLWJYCDAXUIBK-UHFFFAOYSA-N
198234
C1=CC(=CC=C1CCNC(=O)C2=CC=C(C=C2)Cl)O
C15H14ClNO2
MFCD00218513
4-chloro-n-2-4-hydroxyphenyl ethyl benzamide, 4-chloro-n-4-hydroxyphenethyl benzamide, benzamide, 4-chloro-n-2-4-hydroxyphenyl ethyl, benzamide, p-chloro-n-2-p-hydroxyphenyl ethyl, bez-da, n-4-chlorobenzoyl tyramine, n-4-chlorobenzoyl-tyramine, p-chloro-n-2-p-hydroxyphenyl ethyl benzamide
4-chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 255-566-3

RTECSNumber : CV2450481

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents