missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-N-(2,4-dichlorophenyl)butyramide, 97%

Chemical Identifiers

CAS 457618-46-1
Molecular Formula C10H10Cl3NO
Molecular Weight (g/mol) 266.546
MDL Number MFCD03361724
InChI Key SPTDUZNBZNYZFE-UHFFFAOYSA-N
Synonym 4-chloro-n-2,4-dichlorophenyl butanamide, 4-chloro-n-2,4-dichlorophenyl butyramide
PubChem CID 4396570
IUPAC Name 4-chloro-N-(2,4-dichlorophenyl)butanamide
SMILES C1=CC(=C(C=C1Cl)Cl)NC(=O)CCCCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59853MD View Documents Alfa Aesar™
H59853MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5985303 View Documents Alfa Aesar™
H5985303
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

457618-46-1
266.546
SPTDUZNBZNYZFE-UHFFFAOYSA-N
4396570
C1=CC(=C(C=C1Cl)Cl)NC(=O)CCCCl
C10H10Cl3NO
MFCD03361724
4-chloro-n-2,4-dichlorophenyl butanamide, 4-chloro-n-2,4-dichlorophenyl butyramide
4-chloro-N-(2,4-dichlorophenyl)butanamide

Specifications

250mg
C10H10Cl3NO
4-chloro-n-2,4-dichlorophenyl butanamide, 4-chloro-n-2,4-dichlorophenyl butyramide
C1=CC(=C(C=C1Cl)Cl)NC(=O)CCCCl
266.546
266.55
4-Chloro-N-(2,4-dichlorophenyl)butyramide
457618-46-1
MFCD03361724
SPTDUZNBZNYZFE-UHFFFAOYSA-N
4-chloro-N-(2,4-dichlorophenyl)butanamide
4396570
97%