Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-m-xylene, 97%

$76.39 - $280.26

Chemical Identifiers

Chemical Name or Material 4-Chloro-m-xylene
CAS 95-66-9
Molecular Formula C8H9Cl
Formula Weight 140.61
MDL Number MFCD00060644
Synonym 1-Chloro-2,4-dimethylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2127509

  SDS
alfa aesar™
B2127509
10g Each for $76.39

AAB2127518

  SDS
alfa aesar™
B2127518
50g Each for $280.26
Description & Specifications

Specifications

Formula Weight 140.61
Health Hazard 1 Warning
MDL Number MFCD00060644
Chemical Name or Material 4-Chloro-m-xylene
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C8H9Cl
CAS 95-66-9
Synonym 1-Chloro-2,4-dimethylbenzene
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 10g
Assay 97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.