Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-DL-phenylalanine ethyl ester hydrochloride, 98%

$26.81 - $26.81

Chemical Identifiers

Chemical Name or Material 4-Chloro-DL-phenylalanine ethyl ester hydrochloride
CAS 52031-05-7
Molecular Formula C11H14ClNO2·HCl
Formula Weight 264.15
MDL Number MFCD00012531
Beilstein 3574655
Synonym H-Phe(4-Cl)-OEt·HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1279803

  SDS
alfa aesar™
L1279803
1g Each for $26.81
Description & Specifications

Specifications

Formula Weight 264.15
Chemical Name or Material 4-Chloro-DL-phenylalanine ethyl ester hydrochloride
Synonym H-Phe(4-Cl)-OEt·HCl
MDL Number MFCD00012531
Molecular Formula C11H14ClNO2·HCl
Assay 98%
Melting Point 166° to 168°C
Beilstein 3574655
Quantity 1g
CAS 52031-05-7