Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-D-phenylalanine methyl ester hydrochloride, 98%

Chemical Identifiers

Chemical Name or Material 4-Chloro-D-phenylalanine methyl ester hydrochloride
CAS 33965-47-8
Formula Weight 250.12
MDL Number MFCD00236920
Synonym H-D-Phe(4-Cl)-Ome.HCl; (R)-Methyl 2-amino-3-(4-chlorophenyl)propionate hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6387303

  SDS
alfa aesar™
H6387303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6387306

  SDS
alfa aesar™
H6387306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63873MD

  SDS
alfa aesar™
H63873MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 250.12
Chemical Name or Material 4-Chloro-D-phenylalanine methyl ester hydrochloride
Physical Form Powder
Assay 98%
MDL Number MFCD00236920
CAS 33965-47-8
Synonym H-D-Phe(4-Cl)-Ome.HCl; (R)-Methyl 2-amino-3-(4-chlorophenyl)propionate hydrochloride
Color White
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C