missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-D-phenylalanine methyl ester hydrochloride, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6387303

 View more versions of this product

Catalog No. AAH6387303



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Chloro-D-phenylalanine methyl ester hydrochloride
White
MFCD00236920
GCBCWTWQAFLKJG-UHFFFAOYNA-N
methyl 2-amino-3-(4-chlorophenyl)propanoate hydrochloride
24720899
98%
Powder
33965-47-8
C10H13Cl2NO2
4-chloro-d-phenylalanine methyl ester hydrochloride, h-d-phe 4-cl ome.hcl, 4-chloro-d-phe-ome hcl, unii-k88egn008p, r-methyl 2-amino-3-4-chlorophenyl propanoate hydrochloride, d-methyl 4-chlorophenylalaninate hydrochloride, d-phenylalanine, 4-chloro-, methyl ester, hydrochloride, methyl 2r-2-amino-3-4-chlorophenyl propanoate hydrochloride, 4-chloro-d-phe-omecl, 4-chloro-d-phe-ome*hcl
Cl.COC(=O)C(N)CC1=CC=C(Cl)C=C1
250.12
250.12
1 g

Chemical Identifiers

33965-47-8
250.12
GCBCWTWQAFLKJG-UHFFFAOYNA-N
24720899
C10H13Cl2NO2
MFCD00236920
4-chloro-d-phenylalanine methyl ester hydrochloride, h-d-phe 4-cl ome.hcl, 4-chloro-d-phe-ome hcl, unii-k88egn008p, r-methyl 2-amino-3-4-chlorophenyl propanoate hydrochloride, d-methyl 4-chlorophenylalaninate hydrochloride, d-phenylalanine, 4-chloro-, methyl ester, hydrochloride, methyl 2r-2-amino-3-4-chlorophenyl propanoate hydrochloride, 4-chloro-d-phe-omecl, 4-chloro-d-phe-ome*hcl
Cl.COC(=O)C(N)CC1=CC=C(Cl)C=C1
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions