missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-D-phenylalanine methyl ester hydrochloride, 98%

Manufacturer:  Alfa Aesar™ H6387306

 View more versions of this product

Catalog No. AAH6387306



Specifications

Specifications

4-Chloro-D-phenylalanine methyl ester hydrochloride
C10H13Cl2NO2
4-chloro-d-phenylalanine methyl ester hydrochloride, h-d-phe 4-cl ome.hcl, 4-chloro-d-phe-ome hcl, unii-k88egn008p, r-methyl 2-amino-3-4-chlorophenyl propanoate hydrochloride, d-methyl 4-chlorophenylalaninate hydrochloride, d-phenylalanine, 4-chloro-, methyl ester, hydrochloride, methyl 2r-2-amino-3-4-chlorophenyl propanoate hydrochloride, 4-chloro-d-phe-omecl, 4-chloro-d-phe-ome*hcl
COC(=O)C(CC1=CC=C(C=C1)Cl)N.Cl
250.119
250.12
98%
5g
33965-47-8
MFCD00236920
GCBCWTWQAFLKJG-SBSPUUFOSA-N
methyl (2R)-2-amino-3-(4-chlorophenyl)propanoate;hydrochloride
24720899
Powder
White

Chemical Identifiers

33965-47-8
250.119
GCBCWTWQAFLKJG-SBSPUUFOSA-N
24720899
COC(=O)C(CC1=CC=C(C=C1)Cl)N.Cl
C10H13Cl2NO2
MFCD00236920
4-chloro-d-phenylalanine methyl ester hydrochloride, h-d-phe 4-cl ome.hcl, 4-chloro-d-phe-ome hcl, unii-k88egn008p, r-methyl 2-amino-3-4-chlorophenyl propanoate hydrochloride, d-methyl 4-chlorophenylalaninate hydrochloride, d-phenylalanine, 4-chloro-, methyl ester, hydrochloride, methyl 2r-2-amino-3-4-chlorophenyl propanoate hydrochloride, 4-chloro-d-phe-omecl, 4-chloro-d-phe-ome*hcl
methyl (2R)-2-amino-3-(4-chlorophenyl)propanoate;hydrochloride
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents