Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-beta-styrylboronic acid pinacol ester, 97%

$67.50 - $271.22

Chemical Identifiers

Chemical Name or Material 4-Chloro-beta-styrylboronic acid pinacol ester
CAS 223919-54-8
Molecular Formula C14H18BClO2
Formula Weight 264.56
MDL Number MFCD15144834
Beilstein 8260384
Synonym 2-[2-(4-Chlorophenyl)vinyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5169203

  SDS
alfa aesar™
H5169203
1g Each for $67.50

AAH5169206

  SDS
alfa aesar™
H5169206
5g Each for $271.22
Description & Specifications

Specifications

Formula Weight 264.56
CAS 223919-54-8
Assay 97%
MDL Number MFCD15144834
Synonym 2-[2-(4-Chlorophenyl)vinyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Beilstein 8260384
Quantity 1g
Chemical Name or Material 4-Chloro-beta-styrylboronic acid pinacol ester
Molecular Formula C14H18BClO2