Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-alpha-toluenesulfonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 4-Chloro-alpha-toluenesulfonyl chloride
CAS 6966-45-6
Molecular Formula C7H6Cl2O2S
Formula Weight 225.1
MDL Number MFCD01631927
UN Number UN3261
Synonym 4-Chlorobenzylsulfonyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2601703

  SDS
alfa aesar™
H2601703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2601706

  SDS
alfa aesar™
H2601706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.1
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
UN Number UN3261
Health Hazard 1 Danger
Melting Point 93° to 96°C
Assay 97%
CAS 6966-45-6
Chemical Name or Material 4-Chloro-alpha-toluenesulfonyl chloride
Synonym 4-Chlorobenzylsulfonyl chloride
Molecular Formula C7H6Cl2O2S
Quantity 1g
MDL Number MFCD01631927
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.