missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-alpha-methylstyrene (stabilized with TBC), TCI America™

$51.95 - $154.31

Chemical Identifiers

CAS 1712-70-5
Molecular Formula C9H9Cl
Molecular Weight (g/mol) 152.621
MDL Number MFCD00000630
InChI Key WQDGTJOEMPEHHL-UHFFFAOYSA-N
Synonym 4-chloro-alpha-methylstyrene, 4-chloroisopropenylbenzene, benzene, 1-chloro-4-1-methylethenyl, 1-chloro-4-prop-1-en-2-yl benzene, 4-isopropenyl-chlorobenzene, p-chloro-alpha-methylstyrene, 4-chloro-a-methylstyrene, 4-isopropenylchlorobenzene, 4-chloro-.alpha.-methylstyrene, acmc-1bnhn
PubChem CID 74381
IUPAC Name 1-chloro-4-prop-1-en-2-ylbenzene
SMILES CC(=C)C1=CC=C(C=C1)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C021025G View Documents TCI America
C021025G
25g Each for $154.31
 
C02105G View Documents TCI America
C02105G
5g Each for $51.95
 
Specifications

Chemical Identifiers

1712-70-5
152.621
WQDGTJOEMPEHHL-UHFFFAOYSA-N
74381
CC(=C)C1=CC=C(C=C1)Cl
C9H9Cl
MFCD00000630
4-chloro-alpha-methylstyrene, 4-chloroisopropenylbenzene, benzene, 1-chloro-4-1-methylethenyl, 1-chloro-4-prop-1-en-2-yl benzene, 4-isopropenyl-chlorobenzene, p-chloro-alpha-methylstyrene, 4-chloro-a-methylstyrene, 4-isopropenylchlorobenzene, 4-chloro-.alpha.-methylstyrene, acmc-1bnhn
1-chloro-4-prop-1-en-2-ylbenzene

Specifications

207°C
25g
C9H9Cl
3082
WQDGTJOEMPEHHL-UHFFFAOYSA-N
1-chloro-4-prop-1-en-2-ylbenzene
74381
Clear Liquid at 20°C
4-Chloro-alpha-methylstyrene (stabilized with TBC)
Yellow
1712-70-5
MFCD00000630
4-chloro-alpha-methylstyrene, 4-chloroisopropenylbenzene, benzene, 1-chloro-4-1-methylethenyl, 1-chloro-4-prop-1-en-2-yl benzene, 4-isopropenyl-chlorobenzene, p-chloro-alpha-methylstyrene, 4-chloro-a-methylstyrene, 4-isopropenylchlorobenzene, 4-chloro-.alpha.-methylstyrene, acmc-1bnhn
CC(=C)C1=CC=C(C=C1)Cl
152.621
152.62
≥97.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0043&(9)-0452

TSCA : No

Recommended Storage : Refrigerator