missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-chloro-alpha-methylstyrene, 96%, ACROS Organics™

$466.31 - $466.31

Chemical Identifiers

CAS 1712-70-5
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C9H9Cl
Linear Formula H2C=C(CH3)C6H4Cl
Formula Weight 152.62
MDL Number MFCD00000630
Synonym 2-(4-Chlorophenyl)propene
Chemical Name or Material 4-Chloro-α-methylstyrene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC109521000 SDS Thermo Fisher Scientific
109521000
Glass bottle 100g Each for $466.31
Specifications Description & Specifications

Chemical Identifiers

1712-70-5
100.0
C9H9Cl
152.62
2-(4-Chlorophenyl)propene
≥95%
95.0
96%
H2C=C(CH3)C6H4Cl
MFCD00000630
4-Chloro-α-methylstyrene

Specifications

1712-70-5
Authentic
Glass bottle
100g
Liquid
C9H9Cl
152.62
03, 611G
4-Chloro-α-methylstyrene
95.0
100.0
1.5530 to 1.5570 (20°C, 589nm)
Clear colorless to pale yellow
96%
H2C=C(CH3)C6H4Cl
MFCD00000630
2-(4-Chlorophenyl)propene
≥95%