Please login to your online account to display your discounted pricing

4-chloro-alpha-methylstyrene, 96%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  109521000

 View more versions of this product

Catalog No. AC109521000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥95%
Formula Weight 152.62
Physical Form Liquid
Quantity 100g
Color Clear colorless to pale yellow
Beilstein 03, 611G
CAS Max % 100.0
CAS Min % 95.0
Synonym 2-(4-Chlorophenyl)propene
Refractive Index 1.5530 to 1.5570 (20°C, 589nm)
Molecular Formula C9H9Cl
CAS 1712-70-5
Assay Percent Range 96%
Chemical Name or Material 4-Chloro-α-methylstyrene
MDL Number MFCD00000630
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula H2C=C(CH3)C6H4Cl

Specifications

Chemical Name or Material 4-Chloro-α-methylstyrene
CAS 1712-70-5
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C9H9Cl
Linear Formula H2C=C(CH3)C6H4Cl
Formula Weight 152.62
MDL Number MFCD00000630
Synonym 2-(4-Chlorophenyl)propene