Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-alpha-methylstyrene, 90+%, stab. with ca 100ppm 4-tert-butylcatechol

$61.88 - $455.62

Chemical Identifiers

Chemical Name or Material 4-Chloro-alpha-methylstyrene
CAS 1712-70-5
Name Note stabilized with ≈100ppm 4-tert-butylcatechol
Molecular Formula C9H9Cl
Formula Weight 152.62
MDL Number MFCD00000630
Beilstein 1855073
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1491309

 
alfa aesar™
A1491309
10g Each for $61.88

AAA1491318

 
alfa aesar™
A1491318
50g Each for $266.30

AAA1491322

 
alfa aesar™
A1491322
100g Each for $455.62
Description & Specifications

Specifications

Formula Weight 152.62
Boiling Point 205° to 207°C
Chemical Name or Material 4-Chloro-alpha-methylstyrene
Name Note stabilized with ≈100ppm 4-tert-butylcatechol
Assay ≥90%
Molecular Formula C9H9Cl
MDL Number MFCD00000630
CAS 1712-70-5
Beilstein 1855073
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Flash Point 73°C (163°F)
Quantity 10g

Applications
4-Chloro-alpha-methylstyrene is used to produce 3-bromo-2-(4-chlorophenyl)propene at the temperature of 20°C. And it is also used as an organic intermediate.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

2° to 8°C