Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-8-(trifluoromethyl)quinoline, 98%

$75.88 - $75.88

Chemical Identifiers

Chemical Name or Material 4-Chloro-8-(trifluoromethyl)quinoline
CAS 23779-97-7
Molecular Formula C10H5ClF3N
Formula Weight 231.61
MDL Number MFCD00134578
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1730103

 
alfa aesar™
A1730103
1g Each for $75.88
Description & Specifications

Specifications

Formula Weight 231.61
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Chloro-8-(trifluoromethyl)quinoline
Assay 98%
Molecular Formula C10H5ClF3N
MDL Number MFCD00134578
CAS 23779-97-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 80° to 82°C
Quantity 1g

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Low solubility in water.

Notes
Store at room temperature. Keep away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.