Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-8-methoxy-2-methylquinoline, 95%

$171.77 - $747.35

Chemical Identifiers

Chemical Name or Material 4-Chloro-8-methoxy-2-methylquinoline
CAS 64951-58-2
Molecular Formula C11H10ClNO
Formula Weight 207.66
MDL Number MFCD00272401
Synonym 4-Chloro-8-methoxyquinaldine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5055903

  SDS
alfa aesar™
H5055903
1g Each for $171.77

AAH5055906

  SDS
alfa aesar™
H5055906
5g Each for $747.35
Description & Specifications

Specifications

Formula Weight 207.66
Chemical Name or Material 4-Chloro-8-methoxy-2-methylquinoline
Assay 95%
Molecular Formula C11H10ClNO
MDL Number MFCD00272401
CAS 64951-58-2
Synonym 4-Chloro-8-methoxyquinaldine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 89° to 93°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.