Please login to your online account to display your discounted pricing

4-chloro-7-methylthieno[3,2-d]pyrimidine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-Chloro-7-methylthieno[3, 2-d]pyrimidine
CAS 175137-21-0
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H5ClN2S
Formula Weight 184.65
MDL Number MFCD00205202
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

GK01634EA

 
fisher scientific company
GK01634EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 184.65
Quantity 10g
Assay 97%
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H5ClN2S
MDL Number MFCD00205202
CAS 175137-21-0
CAS Min % 96.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Packaging Amber glass bottle
Chemical Name or Material 4-Chloro-7-methylthieno[3, 2-d]pyrimidine
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.