Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-7-fluoro-2-methylquinoline, 95%

$188.35 - $831.73

Chemical Identifiers

Chemical Name or Material 4-Chloro-7-fluoro-2-methylquinoline
CAS 18529-04-9
Molecular Formula C10H7ClFN
Formula Weight 195.63
MDL Number MFCD00272333
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5055403

  SDS
alfa aesar™
H5055403
1g Each for $188.35

AAH5055406

  SDS
alfa aesar™
H5055406
5g Each for $831.73
Description & Specifications

Specifications

Formula Weight 195.63
Chemical Name or Material 4-Chloro-7-fluoro-2-methylquinoline
Assay 95%
Molecular Formula C10H7ClFN
MDL Number MFCD00272333
CAS 18529-04-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
4-Chloro-7-fluoro-2-methylquinoline is used as pharmaceutical intermediates.

Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.