missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-6-methylquinoline, 97%

$78.96 - $251.36

Chemical Identifiers

CAS 18436-71-0
Molecular Formula C10H8ClN
Molecular Weight (g/mol) 177.631
MDL Number MFCD02684204
InChI Key HZWWPOQFLMUYOX-UHFFFAOYSA-N
Synonym quinoline, 4-chloro-6-methyl, 4-chloro-6-methyl-quinoline, acmc-1c6ji, ksc497i2j, 4-chloranyl-6-methyl-quinoline
PubChem CID 824624
IUPAC Name 4-chloro-6-methylquinoline
SMILES CC1=CC2=C(C=CN=C2C=C1)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59890MD View Documents Alfa Aesar™
H59890MD
250mg Each for $78.96
 
AAH5989003 View Documents Alfa Aesar™
H5989003
1g Each for $251.36
 
Specifications

Chemical Identifiers

18436-71-0
177.631
HZWWPOQFLMUYOX-UHFFFAOYSA-N
824624
CC1=CC2=C(C=CN=C2C=C1)Cl
C10H8ClN
MFCD02684204
quinoline, 4-chloro-6-methyl, 4-chloro-6-methyl-quinoline, acmc-1c6ji, ksc497i2j, 4-chloranyl-6-methyl-quinoline
4-chloro-6-methylquinoline

Specifications

250mg
C10H8ClN
quinoline, 4-chloro-6-methyl, 4-chloro-6-methyl-quinoline, acmc-1c6ji, ksc497i2j, 4-chloranyl-6-methyl-quinoline
CC1=CC2=C(C=CN=C2C=C1)Cl
177.631
177.63
4-Chloro-6-methylquinoline
18436-71-0
MFCD02684204
HZWWPOQFLMUYOX-UHFFFAOYSA-N
4-chloro-6-methylquinoline
824624
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning