missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chloro-6-methylpyrimidine, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC454122500
Specifications
4-Chloro-6-methylpyrimidine | |
250mg | |
94% min. (GC) | |
4-chloro-6-methyl-1,3-diazine, 4-chloro-6-methyl-pyrimidine, 4-methyl-6-chloro pyrimidine, 4-methyl-6-chloropyrimidine, 6-chloro-4-methylpyrimidine, 6-methyl-4-chloropyrimidin, ksc269g3h, pubchem13240, pyrimidine, 4-chloro-6-methyl, pyrimidine, 4-chloro-6-methyl-6ci,7ci,8ci,9ci | |
CC1=CC(=NC=N1)Cl | |
128.56 | |
128.56 |
34°C to 36°C | |
3435-25-4 | |
C5H5ClN2 | |
MVAXKFAQKTWRAH-UHFFFAOYSA-N | |
4-chloro-6-methylpyrimidine | |
581796 | |
95% |
Chemical Identifiers
3435-25-4 | |
128.56 | |
4-chloro-6-methyl-1,3-diazine, 4-chloro-6-methyl-pyrimidine, 4-methyl-6-chloro pyrimidine, 4-methyl-6-chloropyrimidine, 6-chloro-4-methylpyrimidine, 6-methyl-4-chloropyrimidin, ksc269g3h, pubchem13240, pyrimidine, 4-chloro-6-methyl, pyrimidine, 4-chloro-6-methyl-6ci,7ci,8ci,9ci | |
4-chloro-6-methylpyrimidine |
C5H5ClN2 | |
MVAXKFAQKTWRAH-UHFFFAOYSA-N | |
581796 | |
CC1=CC(=NC=N1)Cl |
Safety and Handling
GHS Signal Word: Warning