missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-6-methyl-2-phenylquinoline

$244.74 - $1,074.42

Chemical Identifiers

CAS 18618-02-5
Molecular Formula C16H12ClN
Formula Weight 253.73
MDL Number MFCD02179777
Chemical Name or Material 4-Chloro-6-methyl-2-phenylquinoline
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5056103 SDS Alfa Aesar™
H5056103
1g Each for $244.74
AAH5056106 SDS Alfa Aesar™
H5056106
5g Each for $1,074.42
Specifications Description & Specifications

Chemical Identifiers

18618-02-5
253.73
4-Chloro-6-methyl-2-phenylquinoline
C16H12ClN
MFCD02179777

Specifications

1g
C16H12ClN
MFCD02179777
18618-02-5
253.73
4-Chloro-6-methyl-2-phenylquinoline

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning