Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-6-methyl-1,3-phenylene diisocyanate, 98%

Chemical Identifiers

Chemical Name or Material 4-Chloro-6-methyl-1,3-phenylene diisocyanate
CAS 15166-26-4
Molecular Formula C9H5ClN2O2
Formula Weight 208.61
MDL Number MFCD00013846
UN Number UN2811
Beilstein 2973097
Synonym 5-Chloro-2,4-toluene diisocyanate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1871203

 
alfa aesar™
L1871203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1871206

 
alfa aesar™
L1871206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 208.61
Odor Irritating
Assay 98%
Molecular Formula C9H5ClN2O2
MDL Number MFCD00013846
CAS 15166-26-4
Synonym 5-Chloro-2,4-toluene diisocyanate
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Chemical Name or Material 4-Chloro-6-methyl-1,3-phenylene diisocyanate
UN Number UN2811
Beilstein 2973097
Health Hazard 1 Danger
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 61° to 63°C
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C