Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-6-methoxymethyl-2-phenylpyrimidine

$122.05 - $424.91

Chemical Identifiers

Chemical Name or Material 4-Chloro-6-methoxymethyl-2-phenylpyrimidine
CAS 325685-59-4
Molecular Formula C12H11ClN2O
Formula Weight 234.69
MDL Number MFCD00794769
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5053003

  SDS
alfa aesar™
H5053003
1g Each for $424.91

AAH50530MD

  SDS
alfa aesar™
H50530MD
250mg Each for $122.05
Description & Specifications

Specifications

Formula Weight 234.69
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Chemical Name or Material 4-Chloro-6-methoxymethyl-2-phenylpyrimidine
Molecular Formula C12H11ClN2O
MDL Number MFCD00794769
CAS 325685-59-4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.