missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-6-iodoquinazoline, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC451030010

 View more versions of this product

Catalog No. AC451030010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Chloro-6-iodoquinazoline
175.0°C to 177.0°C
98556-31-1
C8H4ClIN2
BDAIUOPDSRAOKI-UHFFFAOYSA-N
4-chloro-6-iodoquinazoline
11173809
97%
Glass Bottle
1g
96% min. (HPLC)
4-chloro-6-iodo quinazoline, 4-chloro-6-iodo-quinazoline, 4-chloro-6-iodo-qulinazoline, 4chloro-6-iodoquinazoline, 6-iodine-4-chloroquine thiazole moiety, 6-iodo-4-chloroquinazoline, 6-iodo4-chloroquinazoline, pubchem23075, quinazoline, 4-chloro-6-iodo
C1=CC2=C(C=C1I)C(=NC=N2)Cl
290.49
290.49

Chemical Identifiers

98556-31-1
290.49
4-chloro-6-iodo quinazoline, 4-chloro-6-iodo-quinazoline, 4-chloro-6-iodo-qulinazoline, 4chloro-6-iodoquinazoline, 6-iodine-4-chloroquine thiazole moiety, 6-iodo-4-chloroquinazoline, 6-iodo4-chloroquinazoline, pubchem23075, quinazoline, 4-chloro-6-iodo
4-chloro-6-iodoquinazoline
C8H4ClIN2
BDAIUOPDSRAOKI-UHFFFAOYSA-N
11173809
C1=CC2=C(C=C1I)C(=NC=N2)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents