missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-6-iodoquinazoline, 97%, ACROS Organics™

Chemical Identifiers

CAS 98556-31-1
Molecular Formula C8H4ClIN2
Molecular Weight (g/mol) 290.49
InChI Key BDAIUOPDSRAOKI-UHFFFAOYSA-N
Synonym 4-chloro-6-iodo quinazoline, 4-chloro-6-iodo-quinazoline, 4-chloro-6-iodo-qulinazoline, 4chloro-6-iodoquinazoline, 6-iodine-4-chloroquine thiazole moiety, 6-iodo-4-chloroquinazoline, 6-iodo4-chloroquinazoline, pubchem23075, quinazoline, 4-chloro-6-iodo
PubChem CID 11173809
IUPAC Name 4-chloro-6-iodoquinazoline
SMILES C1=CC2=C(C=C1I)C(=NC=N2)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC451030010
SDS View Documents
Thermo Fisher Scientific
AC451030010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC451030050
SDS View Documents
Thermo Fisher Scientific
AC451030050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

98556-31-1
290.49
4-chloro-6-iodo quinazoline, 4-chloro-6-iodo-quinazoline, 4-chloro-6-iodo-qulinazoline, 4chloro-6-iodoquinazoline, 6-iodine-4-chloroquine thiazole moiety, 6-iodo-4-chloroquinazoline, 6-iodo4-chloroquinazoline, pubchem23075, quinazoline, 4-chloro-6-iodo
4-chloro-6-iodoquinazoline
C8H4ClIN2
BDAIUOPDSRAOKI-UHFFFAOYSA-N
11173809
C1=CC2=C(C=C1I)C(=NC=N2)Cl

Specifications

Glass Bottle
1g
96% min. (HPLC)
4-chloro-6-iodo quinazoline, 4-chloro-6-iodo-quinazoline, 4-chloro-6-iodo-qulinazoline, 4chloro-6-iodoquinazoline, 6-iodine-4-chloroquine thiazole moiety, 6-iodo-4-chloroquinazoline, 6-iodo4-chloroquinazoline, pubchem23075, quinazoline, 4-chloro-6-iodo
C1=CC2=C(C=C1I)C(=NC=N2)Cl
290.49
290.49
4-Chloro-6-iodoquinazoline
175.0°C to 177.0°C
98556-31-1
C8H4ClIN2
BDAIUOPDSRAOKI-UHFFFAOYSA-N
4-chloro-6-iodoquinazoline
11173809
97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning