missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-6-fluoroquinoline

$90.36 - $309.66

Chemical Identifiers

CAS 391-77-5
Molecular Formula C9H5ClFN
Formula Weight 181.6
MDL Number MFCD00278783
Chemical Name or Material 4-Chloro-6-fluoroquinoline
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5054003 SDS Alfa Aesar™
H5054003
1g Each for $309.66
AAH50540MD SDS Alfa Aesar™
H50540MD
250mg Each for $90.36
Specifications Description & Specifications

Chemical Identifiers

391-77-5
181.6
4-Chloro-6-fluoroquinoline
C9H5ClFN
MFCD00278783

Specifications

1g
C9H5ClFN
MFCD00278783
391-77-5
181.6
4-Chloro-6-fluoroquinoline

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning