Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-6-fluoro-2-methylquinoline, 96%

$105.58 - $380.20

Chemical Identifiers

Chemical Name or Material 4-Chloro-6-fluoro-2-methylquinoline
CAS 18529-01-6
Molecular Formula C10H7ClFN
Formula Weight 195.63
MDL Number MFCD00272332
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5055303

 
alfa aesar™
H5055303
1g Each for $380.20

AAH50553MD

 
alfa aesar™
H50553MD
250mg Each for $105.58
Description & Specifications

Specifications

Formula Weight 195.63
Chemical Name or Material 4-Chloro-6-fluoro-2-methylquinoline
Assay 96%
MDL Number MFCD00272332
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Molecular Formula C10H7ClFN
CAS 18529-01-6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
4-Chloro-6-fluoro-2-methylquinoline react with 2,6-Dinitro-4-trifluormethylnatriumphenolat, and obtain the 4-(2,6-dinitro-4-trifluoromethyl-phenoxy)-6-fluoro-2-methyl-quinoline.

Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.