missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-6-ethyl-5-fluoropyrimidine, 95%

$41.64 - $113.97

Chemical Identifiers

CAS 137234-74-3
Molecular Formula C6H6ClFN2
Molecular Weight (g/mol) 160.576
MDL Number MFCD07782087
InChI Key LKTGVRWVTAJGMS-UHFFFAOYSA-N
Synonym 4-chloro-6-ethyl-5-fluoro-pyrimidine, pyrimidine, 4-chloro-6-ethyl-5-fluoro, 4-chloro-5-fluoro-6-ethylpyrimidine, pubchem7037, acmc-1bzf7, ksc174s6p
PubChem CID 16743445
IUPAC Name 4-chloro-6-ethyl-5-fluoropyrimidine
SMILES CCC1=C(C(=NC=N1)Cl)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6167406 View Documents Alfa Aesar™
H6167406
5g Each for $41.64
 
AAH6167414 View Documents Alfa Aesar™
H6167414
25g Each for $113.97
 
Specifications

Chemical Identifiers

137234-74-3
160.576
LKTGVRWVTAJGMS-UHFFFAOYSA-N
16743445
CCC1=C(C(=NC=N1)Cl)F
C6H6ClFN2
MFCD07782087
4-chloro-6-ethyl-5-fluoro-pyrimidine, pyrimidine, 4-chloro-6-ethyl-5-fluoro, 4-chloro-5-fluoro-6-ethylpyrimidine, pubchem7037, acmc-1bzf7, ksc174s6p
4-chloro-6-ethyl-5-fluoropyrimidine

Specifications

81°C (178°F)
5g
C6H6ClFN2
UN3267
LKTGVRWVTAJGMS-UHFFFAOYSA-N
4-chloro-6-ethyl-5-fluoropyrimidine
16743445
95%
201°C to 211°C
137234-74-3
MFCD07782087
4-chloro-6-ethyl-5-fluoro-pyrimidine, pyrimidine, 4-chloro-6-ethyl-5-fluoro, 4-chloro-5-fluoro-6-ethylpyrimidine, pubchem7037, acmc-1bzf7, ksc174s6p
CCC1=C(C(=NC=N1)Cl)F
160.576
160.58
4-Chloro-6-ethyl-5-fluoropyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.