Learn More
4-Chloro-6-ethyl-5-fluoropyrimidine, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6167406
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Chloro-6-ethyl-5-fluoropyrimidine | |
1.286 | |
81°C (178°F) | |
1.496 | |
5 g | |
4-chloro-6-ethyl-5-fluoro-pyrimidine, pyrimidine, 4-chloro-6-ethyl-5-fluoro, 4-chloro-5-fluoro-6-ethylpyrimidine, pubchem7037, acmc-1bzf7, ksc174s6p | |
CCC1=C(F)C(Cl)=NC=N1 | |
160.58 | |
160.58 |
137234-74-3 | |
201°C to 211°C | |
C6H6ClFN2 | |
MFCD07782087 | |
UN3267 | |
LKTGVRWVTAJGMS-UHFFFAOYSA-N | |
4-chloro-6-ethyl-5-fluoropyrimidine | |
16743445 | |
95% |
Chemical Identifiers
137234-74-3 | |
160.58 | |
LKTGVRWVTAJGMS-UHFFFAOYSA-N | |
16743445 | |
CCC1=C(F)C(Cl)=NC=N1 |
C6H6ClFN2 | |
MFCD07782087 | |
4-chloro-6-ethyl-5-fluoro-pyrimidine, pyrimidine, 4-chloro-6-ethyl-5-fluoro, 4-chloro-5-fluoro-6-ethylpyrimidine, pubchem7037, acmc-1bzf7, ksc174s6p | |
4-chloro-6-ethyl-5-fluoropyrimidine |
Safety and Handling
GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures