missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-chloro-5-nitro-o-phenylenediamine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 350660050

 View more versions of this product

Catalog No. AC350660050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-chloro-5-nitro-o-phenylenediamine
97.5
189.0°C to 193.0°C
Authentic
Glass bottle
MFCD03427594
4-chloro-5-nitro-o-phenylenediamine, 4-chloro-5-nitro-1,2-benzenediamine, 5-chloro-4-nitrobenzene-1,2-diamine, 4-chloro-5-nitro-1,2-phenylenediamine, 4-chloro-5-nitro-benzene-1,2-diamine, 1,2-benzenediamine, 4-chloro-5-nitro
NC1=CC(Cl)=C(C=C1N)[N+]([O-])=O
187.58
187.59
Crystalline Powder
67073-39-6
100.0
Red
97.5% min. (HPLC)
C6H6ClN3O2
5 g
LOQLMWFVXRZASN-UHFFFAOYSA-N
4-chloro-5-nitrobenzene-1,2-diamine
5702618
98%

Chemical Identifiers

67073-39-6
187.58
LOQLMWFVXRZASN-UHFFFAOYSA-N
5702618
NC1=CC(Cl)=C(C=C1N)[N+]([O-])=O
C6H6ClN3O2
MFCD03427594
4-chloro-5-nitro-o-phenylenediamine, 4-chloro-5-nitro-1,2-benzenediamine, 5-chloro-4-nitrobenzene-1,2-diamine, 4-chloro-5-nitro-1,2-phenylenediamine, 4-chloro-5-nitro-benzene-1,2-diamine, 1,2-benzenediamine, 4-chloro-5-nitro
4-chloro-5-nitrobenzene-1,2-diamine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only