Please login to your online account to display your discounted pricing

4-chloro-5-methyl-6-phenylthieno[2,3-d]pyrimidine, 97%, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  AW01194EA

 View more versions of this product

Catalog No. AW01194EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 260.75
Quantity 10g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 4-chloro-5-methyl-6-phenylthieno[2, 3-d]pyrimidine
MDL Number MFCD02179828
Assay Percent Range 97%
Molecular Formula C13H9ClN2S
CAS 306934-78-1
CAS Min % 96.5

Specifications

Chemical Name or Material 4-chloro-5-methyl-6-phenylthieno[2, 3-d]pyrimidine
CAS 306934-78-1
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H9ClN2S
Formula Weight 260.75
MDL Number MFCD02179828