missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-5-methoxy-3(2H)-pyridazinone, TCI America™

Manufacturer:  TCI America C29721G

 View more versions of this product

Catalog No. C29721G



Specifications

Specifications

4-Chloro-5-methoxy-3(2H)-pyridazinone
225°C
63910-43-0
MFCD04971392
COC1=C(C(=O)NN=C1)Cl
160.557
160.56
≥98.0% (GC,T)
White-Yellow
1g
C5H5ClN2O2
SUZFURZANOKZBL-UHFFFAOYSA-N
5-chloro-4-methoxy-1H-pyridazin-6-one
819793
Crystal-Powder at 20°C

Chemical Identifiers

63910-43-0
160.557
SUZFURZANOKZBL-UHFFFAOYSA-N
5-chloro-4-methoxy-1H-pyridazin-6-one
C5H5ClN2O2
MFCD04971392
819793
COC1=C(C(=O)NN=C1)Cl
Safety and Handling

Safety and Handling

TSCA : No

Documents