Please login to your online account to display your discounted pricing

4-chloro-5-(2-thienyl)thieno[2,3-d]pyrimidine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  AW00007DA

 View more versions of this product

Catalog No. AW00007DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 252.75
Quantity 1g
Assay 95%
CAS Max % 100.0
Chemical Name or Material 4-Chloro-5-(2-thienyl)thieno[2, 3-d]pyrimidine
Assay Percent Range 95%
CAS 189681-04-7
Packaging Amber Glass Bottle
MDL Number MFCD00174019
CAS Min % 94.5
Molecular Formula C10H5ClN2S2

Specifications

Chemical Name or Material 4-Chloro-5-(2-thienyl)thieno[2, 3-d]pyrimidine
CAS 189681-04-7
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H5ClN2S2
Formula Weight 252.75
MDL Number MFCD00174019