Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-4'-nitrodiphenyl sulfide, 97%

$245.85 - $245.85

Chemical Identifiers

Chemical Name or Material 4-Chloro-4'-nitrodiphenyl sulfide
CAS 21969-11-9
Molecular Formula C12H8ClNO2S
Formula Weight 265.72g/mol
MDL Number MFCD00043613
Synonym 4-(4-Chlorophenylthio)nitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2055209

 
alfa aesar™
B20552-09
10g Each for $245.85
Description & Specifications

Specifications

Melting Point 83° to 84°C
Quantity 10g
Chemical Name or Material 4-Chloro-4'-nitrodiphenyl sulfide
CAS 21969-11-9
Assay 97%
Molecular Formula C12H8ClNO2S
Formula Weight 265.72g/mol
MDL Number MFCD00043613
Synonym 4-(4-Chlorophenylthio)nitrobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.