Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-4'-methoxybutyrophenone, 97%

$42.32 - $157.00

Chemical Identifiers

Chemical Name or Material 4-Chloro-4'-methoxybutyrophenone
CAS 40877-19-8
Molecular Formula C11H13ClO2
Formula Weight 212.68
MDL Number MFCD00018995
Beilstein 880054
Synonym 3-Chloropropyl 4-methoxyphenyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1545606

 
alfa aesar™
A1545606
5g Each for $42.32

AAA1545614

 
alfa aesar™
A1545614
25g Each for $157.00
Description & Specifications

Specifications

Formula Weight 212.68
Beilstein 880054
Chemical Name or Material 4-Chloro-4'-methoxybutyrophenone
Assay 97%
Molecular Formula C11H13ClO2
MDL Number MFCD00018995
CAS 40877-19-8
Synonym 3-Chloropropyl 4-methoxyphenyl ketone
Flash Point >110°C (230°F)
Boiling Point 159° to 163°C (2mmHg)
Melting Point 30° to 33°C
Quantity 5g