Alfa Aesar™ 4-Chloro-4'-methoxybutyrophenone, 97%
Manufacturer:  Alfa Aesar™ A1545614
 View more versions of this product
Catalog No. AAA1545614
Specifications
Specifications
4-Chloro-4'-methoxybutyrophenone
159°C to 163°C (2mmHg)
25g
C11H13ClO2
880054
NGBTWDPPZFGUAY-UHFFFAOYSA-N
4-chloro-1-(4-methoxyphenyl)butan-1-one
96414
97%
>110°C (230°F)
30°C to 33°C
40877-19-8
MFCD00018995
4-chloro-4'-methoxybutyrophenone, 4-chloro-1-4-methoxyphenyl butan-1-one, acmc-20aml7, gamma-chloro-4'-methoxybutyrophenone, 4-chloro-1-4-methoxyphenyl-1-butanone, 1-butanone,4-chloro-1-4-methoxyphenyl, 4-chloro-1-4-methoxy-phenyl-butan-1-one, 1-butanone, 4-chloro-1-4-methoxyphenyl, 4-chloro-4 inverted exclamation marka-methoxybutyrophenone
COC1=CC=C(C=C1)C(=O)CCCCl
212.673
212.68
Chemical Identifiers
40877-19-8
212.673
NGBTWDPPZFGUAY-UHFFFAOYSA-N
96414
COC1=CC=C(C=C1)C(=O)CCCCl
C11H13ClO2
MFCD00018995
4-chloro-4'-methoxybutyrophenone, 4-chloro-1-4-methoxyphenyl butan-1-one, acmc-20aml7, gamma-chloro-4'-methoxybutyrophenone, 4-chloro-1-4-methoxyphenyl-1-butanone, 1-butanone,4-chloro-1-4-methoxyphenyl, 4-chloro-1-4-methoxy-phenyl-butan-1-one, 1-butanone, 4-chloro-1-4-methoxyphenyl, 4-chloro-4 inverted exclamation marka-methoxybutyrophenone
4-chloro-1-(4-methoxyphenyl)butan-1-one
Documents