missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-4'-hydroxybutyrophenone (Tech.), 95%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 109270250

 View more versions of this product

Catalog No. AC109270250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Crystalline Powder
≥94%
Light beige to gray-green
100.0
Authentic
tech., 95%
MFCD00002362
7150-55-2
95%
198.65
25g
109° to 116°C
94.0
4-Chloro-1-(4-hydroxyphenyl)-1-butanone
4-Chloro-4'-hydroxybutyrophenone
C10H11ClO2
HOC6H4CO(CH2)3Cl

Chemical Identifiers

7150-55-2
100.0
C10H11ClO2
198.65
4-Chloro-1-(4-hydroxyphenyl)-1-butanone
≥94%
94.0
95%
HOC6H4CO(CH2)3Cl
MFCD00002362
4-Chloro-4'-hydroxybutyrophenone, tech., 95%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 230-485-6