Please login to your online account to display your discounted pricing

4-Chloro-4'-hydroxybutyrophenone (Tech.), 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material 4-Chloro-4'-hydroxybutyrophenone
Name Note tech., 95%
CAS 7150-55-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C10H11ClO2
Linear Formula HOC6H4CO(CH2)3Cl
Formula Weight 198.65
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC109270250

  SDS
thermo fisher scientific
109270250
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 198.65
Assay ≥94%
Quantity 25g
Color Light beige to gray-green
EINECS Number 230-485-6
Melting Point 109° to 116°C
CAS Max % 100.0
CAS Min % 94.0
Infrared Spectrum Authentic
Synonym 4-Chloro-1-(4-hydroxyphenyl)-1-butanone
Name Note tech., 95%
Chemical Name or Material 4-Chloro-4'-hydroxybutyrophenone
MDL Number MFCD00002362
Molecular Formula C10H11ClO2
CAS 7150-55-2
Linear Formula HOC6H4CO(CH2)3Cl
Assay Percent Range 95%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.