Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-4'-hydroxybutyrophenone, 95%

$104.42 - $317.93

Chemical Identifiers

Chemical Name or Material 4-Chloro-4'-hydroxybutyrophenone
CAS 7150-55-2
Molecular Formula C10H11ClO2
Formula Weight 198.65
MDL Number MFCD00002362
Synonym 4-Chloro-1-(4-hydroxyphenyl)-1-butanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2554306

  SDS
alfa aesar™
B2554306
5g Each for $104.42

AAB2554314

  SDS
alfa aesar™
B2554314
25g Each for $317.93
Description & Specifications

Specifications

Formula Weight 198.65
Quantity 5g
Chemical Name or Material 4-Chloro-4'-hydroxybutyrophenone
Melting Point 105° to 110°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 95%
Synonym 4-Chloro-1-(4-hydroxyphenyl)-1-butanone
Health Hazard 1 Warning
Molecular Formula C10H11ClO2
CAS 7150-55-2
MDL Number MFCD00002362
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.