missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chloro-4'-hydroxybiphenyl 98.0+%, TCI America™
$232.25 - $784.71
Chemical Identifiers
CAS | 28034-99-3 |
---|---|
Molecular Formula | C12H9ClO |
Molecular Weight (g/mol) | 204.653 |
MDL Number | MFCD00143298 |
InChI Key | ICVFJPSNAUMFCW-UHFFFAOYSA-N |
Synonym | 4-chloro-4'-hydroxybiphenyl, 4-chloro-4'-biphenylol, 4'-chlorobiphenyl-4-ol, 4-4-chlorophenyl phenol, 4-chloro-4-hydroxybiphenyl, 4-biphenylol, 4'-chloro, unii-in91367n9r, 1,1'-biphenyl-4-ol, 4'-chloro, 4'-chloro-4-biphenylol, 4'-chloro-1,1'-biphenyl-4-ol |
PubChem CID | 91589 |
ChEBI | CHEBI:34394 |
IUPAC Name | 4-(4-chlorophenyl)phenol |
SMILES | C1=CC(=CC=C1C2=CC=C(C=C2)Cl)O |
Chemical Identifiers
28034-99-3 | |
204.653 | |
ICVFJPSNAUMFCW-UHFFFAOYSA-N | |
91589 | |
4-(4-chlorophenyl)phenol |
C12H9ClO | |
MFCD00143298 | |
4-chloro-4'-hydroxybiphenyl, 4-chloro-4'-biphenylol, 4'-chlorobiphenyl-4-ol, 4-4-chlorophenyl phenol, 4-chloro-4-hydroxybiphenyl, 4-biphenylol, 4'-chloro, unii-in91367n9r, 1,1'-biphenyl-4-ol, 4'-chloro, 4'-chloro-4-biphenylol, 4'-chloro-1,1'-biphenyl-4-ol | |
CHEBI:34394 | |
C1=CC(=CC=C1C2=CC=C(C=C2)Cl)O |
Specifications
28034-99-3 | |
White | |
MFCD00143298 | |
4-chloro-4'-hydroxybiphenyl, 4-chloro-4'-biphenylol, 4'-chlorobiphenyl-4-ol, 4-4-chlorophenyl phenol, 4-chloro-4-hydroxybiphenyl, 4-biphenylol, 4'-chloro, unii-in91367n9r, 1,1'-biphenyl-4-ol, 4'-chloro, 4'-chloro-4-biphenylol, 4'-chloro-1,1'-biphenyl-4-ol | |
C1=CC(=CC=C1C2=CC=C(C=C2)Cl)O | |
204.653 | |
CHEBI:34394 | |
≥98.0% (T) | |
4-Chloro-4′-hydroxybiphenyl |
146°C | |
C12H9ClO | |
1 g | |
ICVFJPSNAUMFCW-UHFFFAOYSA-N | |
4-(4-chlorophenyl)phenol | |
91589 | |
204.65 | |
Crystalline Powder |
Safety and Handling
TSCA : No