Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-4'-hydroxybenzophenone, 97%

$57.57 - $462.58

Chemical Identifiers

Chemical Name or Material 4-Chloro-4'-hydroxybenzophenone
CAS 42019-78-3
Molecular Formula C13H9ClO2
Formula Weight 232.67
MDL Number MFCD00002357
Beilstein 2049956
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2327009

  SDS
alfa aesar™
B2327009
10g Each for $57.57

AAB2327018

  SDS
alfa aesar™
B2327018
50g Each for $159.72

AAB2327030

  SDS
alfa aesar™
B2327030
250g Each for $462.58
Description & Specifications

Specifications

Formula Weight 232.67
Chemical Name or Material 4-Chloro-4'-hydroxybenzophenone
Assay 97%
Molecular Formula C13H9ClO2
MDL Number MFCD00002357
CAS 42019-78-3
Beilstein 2049956
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 256° to 257°C (13mmHg)
Melting Point 177° to 182°C
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.