Please login to your online account to display your discounted pricing

4-Chloro-4'-fluorobutyrophenone, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Chloro-4'-fluorobutyrophenone
Name Note 97%
CAS 3874-54-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H10ClFO
Linear Formula FC6H4CO(CH2)3Cl
Formula Weight 200.64
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC109231000

  SDS
thermo fisher scientific
AC109231000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC109235000

  SDS
thermo fisher scientific
AC109235000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Oil
Color Slightly yellow-greenish clear
Formula Weight 200.64
Quantity 100g
TSCA TSCA
EINECS Number 223-395-3
CAS Min % 96.0
CAS Max % 100.0
Storage Information The substance may darken on ageing
Synonym 4-Chloro-1-(4-fluorophenyl)-1-butanone
Water 0.2% max. (K.F.)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 97%
Chemical Name or Material 4-Chloro-4'-fluorobutyrophenone
MDL Number MFCD00001007
CAS 3874-54-2
Assay Percent Range 97%
Linear Formula FC6H4CO(CH2)3Cl
Molecular Formula C10H10ClFO
Refractive Index 1.5245 to 1.5265 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Safety and Handling

Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.