Please login to your online account to display your discounted pricing

4-Chloro-4'-fluorobutyrophenone, 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC109231000

 View more versions of this product

Catalog No. AC109231000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Oil
Assay ≥96.0%
Formula Weight 200.64
Color Slightly yellow-greenish clear
Quantity 100g
EINECS Number 223-395-3
TSCA TSCA
CAS Min % 96.0
CAS Max % 100.0
Storage Information The substance may darken on ageing
Synonym 4-Chloro-1-(4-fluorophenyl)-1-butanone
Water 0.2% max. (K.F.)
Chemical Name or Material 4-Chloro-4'-fluorobutyrophenone
Name Note 97%
Assay Percent Range 97%
Molecular Formula C10H10ClFO
Linear Formula FC6H4CO(CH2)3Cl
MDL Number MFCD00001007
CAS 3874-54-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5245 to 1.5265 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 4-Chloro-4'-fluorobutyrophenone
Name Note 97%
CAS 3874-54-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H10ClFO
Linear Formula FC6H4CO(CH2)3Cl
Formula Weight 200.64
MDL Number MFCD00001007
Synonym 4-Chloro-1-(4-fluorophenyl)-1-butanone
Accessories

Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents